Thermo Scientific Chemicals

2-Allyl-N-Fmoc-L-glycine, 95%, Thermo Scientific Chemicals

Catalog number: H52177.03
1 g, Each
Thermo Scientific Chemicals

2-Allyl-N-Fmoc-L-glycine, 95%, Thermo Scientific Chemicals

Catalog number: H52177.03
1 g, Each
Quantity
Catalog number: H52177.03
also known as H52177-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
146549-21-5
IUPAC Name
(2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pent-4-enoate
Molecular Formula
C20H18NO4
InChI Key
YVBLQCANYSFEBN-SFHVURJKSA-M
SMILES
[O-]C(=O)[C@H](CC=C)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Assay (HPLC)
≥94.0%

Description

2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Allyl-N-Fmoc-L-glycine is potentially useful for solid phase peptide synthesis techniques. 2-Allyl-N-Fmoc-L-glycine compound could be useful as an unusual amino acid analog to aid in the deconvolution of protein structure and function. Reagent in the synthesis of a Dicarba-Analog of Octreotide.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Store at 4°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text