N-Boc-4-chloro-D-phenylalanine, 95%, Thermo Scientific Chemicals
N-Boc-4-chloro-D-phenylalanine, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-4-chloro-D-phenylalanine, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 mg
1 g
Catalog number H52181.06
also known as H52181-06
Price (USD)/ Each
250.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
250.00
Add to cart
N-Boc-4-chloro-D-phenylalanine, 95%, Thermo Scientific Chemicals
Catalog numberH52181.06
Price (USD)/ Each
250.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Boc-4-chloro-D-phenylalanine
CAS57292-44-1
Melting Point110°C
Recommended StorageAmbient temperatures
TSCANo
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only