N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%, Thermo Scientific Chemicals
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52188.MD
also known as H52188-MD
Price (USD)/ Each
141.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
141.00
Add to cart
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%, Thermo Scientific Chemicals
Catalog numberH52188.MD
Price (USD)/ Each
141.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester
CAS203854-49-3
Melting Point115°C to 120°C
Recommended StorageAmbient temperatures
TSCANo
View more
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only