N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%, Thermo Scientific Chemicals
N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52191.MD
also known as H52191-MD
Price (USD)/ Each
140.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
140.00
Add to cart
N-Boc-L-beta-glutamic acid 5-benzyl ester, 95%, Thermo Scientific Chemicals
Catalog numberH52191.MD
Price (USD)/ Each
140.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Boc-L-beta-glutamic acid 5-benzyl ester
CAS254101-10-5
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC17H23NO6
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only