Methyl 2-pyridyl ketoxime, 97%
Methyl 2-pyridyl ketoxime, 97%
Methyl 2-pyridyl ketoxime, 97%
Methyl 2-pyridyl ketoxime, 97%
Thermo Scientific Chemicals

Methyl 2-pyridyl ketoxime, 97%

CAS: 1758-54-9 | C7H8N2O | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H52232.06
also known as H52232-06
Price (USD)
166.00
Each
Estimated availability date  Pending  
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
166.00
Each
Add to cart
Specifications
CAS1758-54-9
Chemical Name or MaterialMethyl 2-pyridyl ketoxime
Formula Weight136.15
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Ask our AI about this Product
Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl-2-pyridyl ketoxime is used as colorimetric reagent for rhenium. Methyl-2-pyridyl ketoxime has been proposed as an analytical reagent for the spectrophotometric determination of copper, iron in alkaline aqueous solution and phenyl-2 pyridyl ketoxime.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Richard J. Thompson,; Ronald H. Gore 1, Fred Trusell.Methyl-2-pyridyl ketoxime as a colorimetric reagent for rhenium. Analytica Chimica Acta. 1964 , 31 590-594.
  2. D. Banerjea,; K. K. Tripathi.Spectrophotometric Determination of Iron and Copper with Methyl-2-Pyridyl Ketoxime and Their Simultaneous Determination in Mixtures. Anal. Chem., 1960, 32 (9), 1196-1200.
Have questions about this product? Ask our AI assisted search.
Are the Pro-Detect assay kits compatible with common detergent and salt concentrations used in protein purification/detection protocols?
How do I interpret the gradual disappearance of four lines in the Pro-Detect competition assay kits?
Are the results of Pro-Detect assay kits comparable to western blotting?
Using Pro-Detect assay kits, can I detect the protein of interest in a mixture of proteins such as a crude lysate?
I am using a Pro-Detect assay kit and cannot differentiate banding pattern between control lysates and lysates that are actually expressing proteins of interest. What is the problem?
+Show more FAQs for this product
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
B21X041Certificate of AnalysisDec 07, 2021H52232.06, H52232.03
1 result displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link