Thermo Scientific Chemicals

Hafnium(IV) isopropoxide isopropanol adduct, 99%, Thermo Scientific Chemicals

Catalog number: H52288.14
25 g, Each
Thermo Scientific Chemicals

Hafnium(IV) isopropoxide isopropanol adduct, 99%, Thermo Scientific Chemicals

Catalog number: H52288.14
25 g, Each
Quantity
Catalog number: H52288.14
also known as H52288-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
2171-99-5
IUPAC Name
tetrakis(propan-2-ol) hafnium
Molecular Formula
C12H32HfO4
InChI Key
HMKGKDSPHSNMTM-UHFFFAOYSA-N
SMILES
[Hf].CC(C)O.CC(C)O.CC(C)O.CC(C)O
Assay from Supplier's CofA
≥98.5% (equates to Hafnium: ≥37.2%)
Appearance (Color)
White
Form
Powder or crystals or crystalline powder

Description

Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hafnium(IV) isopropoxide isopropanol adduct is used as organic chemical synthesis intermediate.

Solubility
It reacts with water.

Notes
Moisture sensitive. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text