Thermo Scientific Chemicals

(2-Aminoethoxy)acetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H52289.03
1 g, Each
Thermo Scientific Chemicals

(2-Aminoethoxy)acetic acid, 98%, Thermo Scientific Chemicals

Catalog number: H52289.03
1 g, Each
Quantity
Catalog number: H52289.03
also known as H52289-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10366-71-9
IUPAC Name
2-(2-aminoethoxy)acetic acid
Molecular Formula
C4H9NO3
InChI Key
GNRLUBOJIGSVNT-UHFFFAOYSA-N
SMILES
NCCOCC(O)=O
Appearance (Color)
White to pale cream
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder
Proton NMR
Conforms to structure

Description

Reactant in the synthesis ammonium ion-selective fluoroionophore. Used in the analysis of recognition elements for mitochondrial processing peptidase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis ammonium ion-selective fluoroionophore. Used in the analysis of recognition elements for mitochondrial processing peptidase.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text