2'-Deoxyinosine, 98+%
2'-Deoxyinosine, 98+%
Thermo Scientific Chemicals

2'-Deoxyinosine, 98+%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H52292.MD
also known as H52292-MD
Price (USD)/ Each
20.60
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
20.60
Add to cart
2'-Deoxyinosine, 98+%
Catalog numberH52292.MD
Price (USD)/ Each
20.60
-
Add to cart
Chemical Identifiers
CAS890-38-0
IUPAC Name9-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular FormulaC10H12N4O4
InChI KeyVGONTNSXDCQUGY-YUZWJPFSNA-N
SMILESOC[C@H]1O[C@H](C[C@@H]1O)N1C=NC2=C1N=CNC2=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.0% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only