Thermo Scientific Chemicals

(S)-1,4-Diazabicyclo[4.3.0]nonane, 98+%, Thermo Scientific Chemicals

Catalog number: H52295.MD
250 mg, Each
Thermo Scientific Chemicals

(S)-1,4-Diazabicyclo[4.3.0]nonane, 98+%, Thermo Scientific Chemicals

Catalog number: H52295.MD
250 mg, Each
Quantity
Catalog number: H52295.MD
also known as H52295-MD
Price (USD)
Quantity
-
Have Questions?

Description

(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1,4-Diazabicyclo[4.3.0]nonane is used as stationary phases for the separation of enantiomeric mixtures of products. It is also used as catalysts for the enantioselective synthesis of various organic compounds and drugs.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text