N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52309.03
also known as H52309-03
Price (USD)/ Each
186.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
186.00
Add to cart
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine, 98+%, Thermo Scientific Chemicals
Catalog numberH52309.03
Price (USD)/ Each
186.00
-
Add to cart
Specifications
Chemical Name or MaterialN-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine
CAS51549-39-4
Recommended StorageAmbient temperatures
TSCANo
Molecular FormulaC23H31N5O4Si
View more
N-Benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoyl-5′-O-tert-butyldimethylsilyl-2′-deoxyadenosine is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. S.K.Burley; A.H.J.Wang. Structure of N-benzoyl-5'-O-tert-butyldimethylsilyl-2'-deoxyadenosine monohydrate. Acta Cryst. 1987, C43, 988-990