5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H52330.03
also known as H52330-03
Price (USD)/ Each
533.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
533.00
Add to cart
5-Carbamoylpyridine-3-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals
Catalog numberH52330.03
Price (USD)/ Each
533.00
-
Add to cart
Specifications
Chemical Name or Material5-Carbamoylpyridine-3-boronic acid pinacol ester
CAS1169402-51-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Carbamoylpyridine-3-boronic acid pinacol ester is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only