Thermo Scientific Chemicals

4-Amino-2,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog number: H52356.03
1 g, Each
Thermo Scientific Chemicals

4-Amino-2,5-difluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific Chemicals

Catalog number: H52356.03
1 g, Each
Quantity
Catalog number: H52356.03
also known as H52356-03
Price (USD)
Quantity
-
Have Questions?
Purity
≥95.0% (H NMR)
Appearance (Color)
White
Form
Powder or crystals or crystalline powder
Proton NMR
Conforms

Description

4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-2,5-difluorobenzeneboronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text