1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-(2-Aminoethyl)-4-methylpiperazine is used as pharmaceutical intermediate.
Notes
Air sensitive. Incompatible with oxidizing agents, acids, acid chlorides and acid anhydrides.
RUO – Research Use Only
Gavin W.Collie; Rossella Promontorio; Sonja M.Hampel; Marialuisa Micco; Stephen Neidle; Gary N.Parkinson. Structural basis for telomeric G-Quadruplex targeting by naphthalene diimide ligands. J. Am. Chem. Soc. 2012, 134, (5),2723-2731