Thermo Scientific Chemicals

(+/-)-3-(Boc-amino)pyrrolidine, 97%, Thermo Scientific Chemicals

Catalog number: H52414.03
1 g, Each
Thermo Scientific Chemicals

(+/-)-3-(Boc-amino)pyrrolidine, 97%, Thermo Scientific Chemicals

Catalog number: H52414.03
1 g, Each
Quantity
Catalog number: H52414.03
also known as H52414-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99724-19-3
IUPAC Name
(3R)-3-{[(tert-butoxy)carbonyl]amino}pyrrolidin-1-ium
Molecular Formula
C9H19N2O2
InChI Key
DQQJBEAXSOOCPG-SSDOTTSWSA-O
SMILES
CC(C)(C)OC(=O)N[C@@H]1CC[NH2+]C1
Residue on ignition
0.10% max
Appearance (Color)
White to pale yellow
Assay (GC)
> 96%
Loss on Drying
0.5% max
Proton NMR
Conforms to structure

Description

(±)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-3-(Boc-amino)pyrrolidine is the homochiral starting materials utilised in the synthesis of all of the chiral compounds. Reactant in the peptide activated G-protein coupled receptors test.

Solubility
Soluble in Methanol(almost transparency).

Notes
Air sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text