1-Boc-D-nipecotic acid, 97%, Thermo Scientific Chemicals
1-Boc-D-nipecotic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-Boc-D-nipecotic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H52416.06
also known as H52416-06
Price (USD)/ Each
106.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
106.00
Add to cart
1-Boc-D-nipecotic acid, 97%, Thermo Scientific Chemicals
Catalog numberH52416.06
Price (USD)/ Each
106.00
-
Add to cart
Chemical Identifiers
CAS163438-09-3
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Loss on Drying<0.50%
Optical Rotation-50.0? to -53.0? (2.5% in Methanol)
Assay (unspecified)>96.0%
Optical purity>98.0%
View more
1-Boc-D-nipecotic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-D-nipecotic acid is used as an organic chemical synthesis intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Jean-Luc Fauchere; Peter W.Schiller. Synthesis, characterization and biological properties of five enkephalin-like pentapeptides containing p-nitrophenylalanine. Helvetica Chimica Acta. 1981, 64, (7),2090-2094
  2. Jean-Luc Fauchere; Susanne Pfenninger; Kim Quang Do; Carole Lemieux; Peter W.Schiller. Synthesis and opiate activity in vitro of five new p-nitrophenylalanine4-enkephalin-like peptides. Helvetica Chimica Acta. 1983, 66, (4),1053-1060