(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H52418.03
also known as H52418-03
Price (USD)/ Each
637.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
637.00
Add to cart
(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberH52418.03
Price (USD)/ Each
637.00
-
Add to cart
Specifications
Chemical Name or Material(1R,2R)-2-Aminocyclohexanecarboxylic acid
CAS26685-83-6
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Albrecht Berkessel; Katja Glaubitz; Johann Lex. Enantiomerically pure β-amino acids: A convenient access to both enantiomers of trans-2-aminocyclohexanecarboxylic acid. European Journal of Organic Chemistry. 2002, 2002, (17),2948-2952
  2. Daniel H.Appella; Laurie A.Christianson; Isabella L.Karle; Douglas R.Powell; Samuel H.Gellman. Synthesis and characterization of trans-2-aminocyclohexanecarboxylic acid oligomers: An unnatural helical secondary structure and implications for β-peptide tertiary structure. J. Am. Chem. Soc. 1999, 121, (26),6206-6212