Thermo Scientific Chemicals

(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52418.MD
250 mg, Each
Thermo Scientific Chemicals

(1R,2R)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52418.MD
250 mg, Each
Quantity
Catalog number: H52418.MD
also known as H52418-MD
Price (USD)
Quantity
-
Have Questions?

Description

(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1R,2R)-2-Aminocyclohexanecarboxylic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text