(R)-3-Amino-1-benzylpiperidine, 97%, Thermo Scientific Chemicals
(R)-3-Amino-1-benzylpiperidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-3-Amino-1-benzylpiperidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H52425.06
also known as H52425-06
Price (USD)/ Each
309.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
309.00
Add to cart
(R)-3-Amino-1-benzylpiperidine, 97%, Thermo Scientific Chemicals
Catalog numberH52425.06
Price (USD)/ Each
309.00
-
Add to cart
Specifications
Chemical Name or Material(R)-3-Amino-1-benzylpiperidine
CAS168466-84-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-Amino-1-benzylpiperidine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only
Jacinto Moragues; Jose Prieto; Robert G.W.Spickett; Armando Vega. Rearrangements during the synthesis of substituted 1-benzylpyrrolidines and 3-substituted 1-benzylpiperidines. J. Chem. Soc., Perkin Trans. 1. 1976, (9),938-940