3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52510.03
also known as H52510-03
Price (USD)/ Each
115.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
115.00
Add to cart
3-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH52510.03
Price (USD)/ Each
115.00
-
Add to cart
Chemical Identifiers
CAS505083-04-5
IUPAC Name[3-fluoro-4-(methoxycarbonyl)phenyl]boronic acid
Molecular FormulaC8H8BFO4
InChI KeyYZYGXFXSMDUXJT-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(C=C1F)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only