3-(2-Methoxycarbonylethyl)benzeneboronic Acid, 97%, Thermo Scientific Chemicals
3-(2-Methoxycarbonylethyl)benzeneboronic Acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(2-Methoxycarbonylethyl)benzeneboronic Acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H52542.MD
also known as H52542-MD
Price (USD)/ Each
135.00
-
Add to cart
Quantity:
250 mg
Request bulk or custom format
Price (USD)/ Each
135.00
Add to cart
3-(2-Methoxycarbonylethyl)benzeneboronic Acid, 97%, Thermo Scientific Chemicals
Catalog numberH52542.MD
Price (USD)/ Each
135.00
-
Add to cart
Specifications
Chemical Name or Material3-(2-Methoxycarbonylethyl)benzeneboronic acid
CAS833472-82-5
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(2-Methoxycarbonylethyl)benzeneboronic acid is involved in the preparation of antimalarial and anticancer ethylenediamine inhibitors to protein farnesyltransferase.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
Hast, M. A.; Steven, F.; Christopher, G. C.; Erin, E. P.; Michelle, A. B.; Kasey, R.; Michael, H. G.; Wesley, C. V. V.; Said, M. S.; Andrew, D. H.; Beese, L. S. Structural Basis for Binding and Selectivity of Antimalarial and Anticancer Ethylenediamine Inhibitors to Protein Farnesyltransferase. Chem. Biol. 2009, 16 (2), 181-192.