Thermo Scientific Chemicals

Phenanthrene-9-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52546.06
5 g, Each
Thermo Scientific Chemicals

Phenanthrene-9-boronic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52546.06
5 g, Each
Quantity
Catalog number: H52546.06
also known as H52546-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
68572-87-2
Assay (HPLC)
96.0% min

Description

Phenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenanthrene-9-boronic acid as a reactant is involved in the synthesis of large polycyclic aromatic hydrocarbons (PAHs) and electron rich benzo[g,h,i]perylenes. It is also used for preparation of FLAP protein inhibitors as anti-inflammatory agents and aminoisoquinolinones as PARP-2 selective inhibitors. As a reactant it is involved in homocoupling reactions.

Solubility
Soluble in DMSO.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text