Thermo Scientific™

1-Boc-3-iodoazetidine, 97%, Thermo Scientific™

Catalog number: H52558.06
Each
Thermo Scientific™

1-Boc-3-iodoazetidine, 97%, Thermo Scientific™

Catalog number: H52558.06
Each
Catalog number: H52558.06
also known as H52558-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
254454-54-1
IUPAC Name
tert-butyl 3-iodoazetidine-1-carboxylate
Molecular Formula
C8H14INO2
InChI Key
XPDIKRMPZNLBAC-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CC(I)C1
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
>96.0%
Refractive Index
1.5093 @20?C
Form
Liquid

Description

1-Boc-3-iodoazetidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Boc-3-iodoazetidine is used as an organic chemical synthesis intermediate.

Notes
Light sensitive. Incompatible with oxidizing agents.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text