(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific Chemicals
(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number H52559.06
also known as H52559-06
Price (USD)/ Each
195.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
195.00
Add to cart
(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific Chemicals
Catalog numberH52559.06
Price (USD)/ Each
195.00
-
Add to cart
Chemical Identifiers
CAS140695-85-8
IUPAC Nametert-butyl 3-(hydroxymethyl)piperidine-1-carboxylate
Molecular FormulaC11H21NO3
InChI KeyOJCLHERKFHHUTB-UHFFFAOYNA-N
SMILESCC(C)(C)OC(=O)N1CCCC(CO)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals
Assay (unspecified)>96.0%
Loss on Drying<0.50%
Optical purity>98.0%
View more
(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-(-)-1-Boc-3-(hydroxymethyl)piperidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only