4-Isobutylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
4-Isobutylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isobutylbenzeneboronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52603.03
also known as H52603-03
Price (USD)/ Each
29.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
29.00
Add to cart
4-Isobutylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Catalog numberH52603.03
Price (USD)/ Each
29.00
-
Add to cart
Chemical Identifiers
CAS153624-38-5
IUPAC Name[4-(2-methylpropyl)phenyl]boronic acid
Molecular FormulaC10H15BO2
InChI KeyYZSPHVWALUJQNK-UHFFFAOYSA-N
SMILESCC(C)CC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>97.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only