4-Isopropoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
4-Isopropoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Isopropoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
1 g
5 g
Catalog number H52675.14
also known as H52675-14
Price (USD)/ Each
202.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
202.00
Add to cart
4-Isopropoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH52675.14
Price (USD)/ Each
202.00
-
Add to cart
Chemical Identifiers
CAS153624-46-5
IUPAC Name[4-(propan-2-yloxy)phenyl]boronic acid
Molecular FormulaC9H13BO3
InChI KeyCJUHQADBFQRIMC-UHFFFAOYSA-N
SMILESCC(C)OC1=CC=C(C=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only