Chemical Identifiers
CAS88050-17-3
IUPAC Name(2S,4R)-1-{[(9H-fluoren-9-yl)methoxy]carbonyl}-4-hydroxypyrrolidine-2-carboxylic acid
Molecular FormulaC20H19NO5
InChI KeyGOUUPUICWUFXPM-XIKOKIGWSA-N
SMILESO[C@@H]1C[C@H](N(C1)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View more