Thermo Scientific Chemicals

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 97%, Thermo Scientific Chemicals

Catalog number: H52745.06
5 g, Each
Thermo Scientific Chemicals

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate, 97%, Thermo Scientific Chemicals

Catalog number: H52745.06
5 g, Each
Quantity
Catalog number: H52745.06
also known as H52745-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
680-15-9
IUPAC Name
methyl 2,2-difluoro-2-(fluorosulfonyl)acetate
Molecular Formula
C3H3F3O4S
InChI Key
GQJCAQADCPTHKN-UHFFFAOYSA-N
SMILES
COC(=O)C(F)(F)S(F)(=O)=O
Assay (unspecified)
>96.0%
Appearance (Color)
Colorless to light yellow
Water Content (Karl Fischer Titration)
<0.50%
Form
Liquid

Description

Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 2,2-difluoro-2-(fluorosulfonyl)acetate is a useful reagent for the trifluoromethylation of alkyl halides. It is also broadly applicable reagent for perfluoroalkylations for aryl iodides and bromides.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text