Thermo Scientific Chemicals

(R)-3-(Boc-amino)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52749.03
1 g, Each
Thermo Scientific Chemicals

(R)-3-(Boc-amino)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52749.03
1 g, Each
Quantity
Catalog number: H52749.03
also known as H52749-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
309956-78-3
IUPAC Name
tert-butyl N-[(3R)-piperidin-3-yl]carbamate
Molecular Formula
C10H20N2O2
InChI Key
WUOQXNWMYLFAHT-MRVPVSSYSA-N
SMILES
CC(C)(C)OC(=O)N[C@@H]1CCCNC1
Appearance (Color)
White to pale yellow
Form
Solid
Assay (unspecified)
>96.0%
Loss on Drying
<0.50%
Optical purity
>98.0%

Description

(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(R)-3-(Boc-amino)piperidine is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol and ethanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text