Thermo Scientific Chemicals

(S)-1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52771.03
1 g, Each
Thermo Scientific Chemicals

(S)-1-Boc-3-hydroxypiperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52771.03
1 g, Each
Quantity
Catalog number: H52771.03
also known as H52771-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
143900-44-1
IUPAC Name
tert-butyl 3-hydroxypiperidine-1-carboxylate
Molecular Formula
C10H19NO3
InChI Key
UIJXHKXIOCDSEB-UHFFFAOYNA-N
SMILES
CC(C)(C)OC(=O)N1CCCC(O)C1
Appearance (Color)
White to off-white
Assay (unspecified)
>96.0%
Optical purity
>98.0%
Optical Rotation
+21.0? to +24.0? (5% in MeOH)
Loss on Drying
<0.75%

Description

(S)-1-Boc-3-hydroxypiperidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-1-Boc-3-hydroxypiperidine is used as pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text