Thermo Scientific Chemicals

(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52779.03
1 g, Each
Thermo Scientific Chemicals

(1S,2S)-2-Aminocyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52779.03
1 g, Each
Quantity
Catalog number: H52779.03
also known as H52779-03
Price (USD)
Quantity
-
Have Questions?

Description

(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(1S,2S)-2-Aminocyclohexanecarboxylic acid is used in the preparation of enantiomerically pure beta-amino acids. It is also involved in the synthesis of mono and dihydroxylated derivatives of 2-aminocyclohexanecarboxylic acid by Diels-Alder adduct of ethyl(E)-3-nitroacrylate and furan.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text