Thermo Scientific Chemicals

(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52784.14
25 g, Each
Thermo Scientific Chemicals

(+/-)-1-Boc-3-(hydroxymethyl)piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52784.14
25 g, Each
Quantity
Catalog number: H52784.14
also known as H52784-14
Price (USD)
Quantity
-
Have Questions?

Description

(±)-1-Boc-3-(hydroxymethyl)piperidine is used as a reactant for synthesis of Pim-1 inhibitors, Vasopressin1b receptor antagonists, CXCR4 antagonists as anti-HIV agents, Amide CCR5 antagonist, PSSRI-based inhibitors of S. aureus multidrug efflux pumps and Human GnRH receptor antagonists.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-1-Boc-3-(hydroxymethyl)piperidine is used as a reactant for synthesis of Pim-1 inhibitors, Vasopressin1b receptor antagonists, CXCR4 antagonists as anti-HIV agents, Amide CCR5 antagonist, PSSRI-based inhibitors of S. aureus multidrug efflux pumps and Human GnRH receptor antagonists.

Solubility
Soluble in methanol and dimethylformamide(DMF).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text