Thermo Scientific Chemicals

1-Boc-azetidine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H52794.03
1 g, Each
Thermo Scientific Chemicals

1-Boc-azetidine-3-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H52794.03
1 g, Each
Quantity
Catalog number: H52794.03
also known as H52794-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
177947-96-5
IUPAC Name
tert-butyl 3-formylazetidine-1-carboxylate
Molecular Formula
C9H15NO3
InChI Key
JVQOZRRUGOADSU-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CC(C1)C=O
Refractive Index
1.4590-1.4665 @ 20°C
Form
Liquid
Free acid (titration)
<2.0%
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥96.0%

Description

1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-azetidine-3-carboxaldehyde is used as a starting material in organic synthesis. It is also used as an intermediate in pharmaceuticals.

Notes
Air sensitive. Store in a cool place. Incompatible with water and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text