Thermo Scientific Chemicals

1-Boc-piperidine-4-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H52813.03
1 g, Each
Thermo Scientific Chemicals

1-Boc-piperidine-4-carboxaldehyde, 97%, Thermo Scientific Chemicals

Catalog number: H52813.03
1 g, Each
Quantity
Catalog number: H52813.03
also known as H52813-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137076-22-3
IUPAC Name
tert-butyl 4-formylpiperidine-1-carboxylate
Molecular Formula
C11H19NO3
InChI Key
JYUQEWCJWDGCRX-UHFFFAOYSA-N
SMILES
CC(C)(C)OC(=O)N1CCC(CC1)C=O
Form
Crystals
Appearance (Color)
White to off-white
Assay (unspecified)
>96.0%
Loss on Drying
<0.50%

Description

1-Boc-piperidine-4-carboxaldehyde is used as reactant for synthesis of Pim-1 inhibitors, selective GPR119 agonists for type II diabetes, M-tropic (R5) HIV-1 replication inhibitors, HDAC inhibitors and selective 5-HT6 antagonists. It is also used as reactant for three-component vinylogous Mannich reactions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Boc-piperidine-4-carboxaldehyde is used as reactant for synthesis of Pim-1 inhibitors, selective GPR119 agonists for type II diabetes, M-tropic (R5) HIV-1 replication inhibitors, HDAC inhibitors and selective 5-HT6 antagonists. It is also used as reactant for three-component vinylogous Mannich reactions

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text