Thermo Scientific Chemicals

4-Aminomethyl-1-Boc-piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52819.06
5 g, Each
Thermo Scientific Chemicals

4-Aminomethyl-1-Boc-piperidine, 97%, Thermo Scientific Chemicals

Catalog number: H52819.06
5 g, Each
Quantity
Catalog number: H52819.06
also known as H52819-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
144222-22-0
Assay (unspecified)
>96.0%
Water Content (Karl Fischer Titration)
<0.50%
Solution Test
Freely soluble in Methanol
Appearance (Color)
Colorless
Form
Liquid

Description

4-Aminomethyl-1-Boc-piperidine is used in the preparation biologically active compounds and orally active platelet-activating factor antagonists. It is used as an inhibitor for aspartic acid protease and Kinesin spindle protein. It is an active reactant involved in the enantioselective synthesis of N-alkyl terminal aziridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Aminomethyl-1-Boc-piperidine is used in the preparation biologically active compounds and orally active platelet-activating factor antagonists. It is used as an inhibitor for aspartic acid protease and Kinesin spindle protein. It is an active reactant involved in the enantioselective synthesis of N-alkyl terminal aziridines.

Solubility
Soluble in chloroform and methanol.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text