3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals
3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H52831.03
also known as H52831-03
Price (USD)/ Each
276.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
276.00
Add to cart
3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals
Catalog numberH52831.03
Price (USD)/ Each
276.00
-
Add to cart
Chemical Identifiers
CAS914348-04-2
IUPAC Namebenzyl N-(azetidin-3-yl)carbamate
Molecular FormulaC11H14N2O2
InChI KeyUWOOBFRZDNDUQB-UHFFFAOYSA-N
SMILESO=C(NC1CNC1)OCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)96.0% min
Water Content (Karl Fischer Titration)7% - 8.5%
3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only
Rosario Herranz; Santiago Conde; Piedad Fernandez-Resa; Enrique Arribas. Enantioselective synthesis of (3S)-trans-4-(substituted methyl)monobactams from 2,3-O-isopropylidene-D-glyceraldehyde. J. Chem. Soc., Perkin Trans. 1. 1988, (3),649-655