Thermo Scientific Chemicals

3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals

Catalog number: H52831.06
5 g, Each
Thermo Scientific Chemicals

3-(Benzyloxycarbonylamino)azetidine, 97%, Thermo Scientific Chemicals

Catalog number: H52831.06
5 g, Each
Quantity
Catalog number: H52831.06
also known as H52831-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
914348-04-2
IUPAC Name
benzyl N-(azetidin-3-yl)carbamate
Molecular Formula
C11H14N2O2
InChI Key
UWOOBFRZDNDUQB-UHFFFAOYSA-N
SMILES
O=C(NC1CNC1)OCC1=CC=CC=C1
Assay (GC)
96.0% min
Water Content (Karl Fischer Titration)
7% - 8.5%

Description

3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Benzyloxycarbonylamino)azetidine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text