Thermo Scientific Chemicals

4-Nitrobenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H52848.03
1 g, Each
Thermo Scientific Chemicals

4-Nitrobenzeneboronic acid pinacol ester, 98%, Thermo Scientific Chemicals

Catalog number: H52848.03
1 g, Each
Quantity
Catalog number: H52848.03
also known as H52848-03
Price (USD)
Quantity
-
Have Questions?

Description

4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Nitrobenzeneboronic acid pinacol ester is a reactant involved in synthesis of aryl azides, arylation of allylic chlorides, synthesis of RNA conjugates, alkoxycarbonylation, synthesis of mTOR and PI3K inhibitors as antitumor agents, electro oxidative synthesis of biaryls.

Solubility
Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text