Thermo Scientific Chemicals

4-Methoxycarbonyl-2-nitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52863.06
5 g, Each
Thermo Scientific Chemicals

4-Methoxycarbonyl-2-nitrobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Catalog number: H52863.06
5 g, Each
Quantity
Catalog number: H52863.06
also known as H52863-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85107-55-7
IUPAC Name
[4-(methoxycarbonyl)-2-nitrophenyl]boronic acid
Molecular Formula
C8H8BNO6
InChI Key
SFEJGHJCGQNFQC-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(B(O)O)C(=C1)[N+]([O-])=O
Assay (HPLC)
>96.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text