Thermo Scientific Chemicals

4-n-Butoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H52865.03
1 g, Each
Thermo Scientific Chemicals

4-n-Butoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H52865.03
1 g, Each
Quantity
Catalog number: H52865.03
also known as H52865-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
105365-51-3
IUPAC Name
(4-butoxyphenyl)boronic acid
Molecular Formula
C10H15BO3
InChI Key
QUPFQMXWFNJUNJ-UHFFFAOYSA-N
SMILES
CCCCOC1=CC=C(C=C1)B(O)O
Assay (HPLC)
>97.5%

Description

Used as a reactant for Suzuki-Miyaura cross-coupling reactions, condensation reactions, synthesis of azobenzene-functionalized compounds. Also used for preparation of highly fluorescent diketopyrrolopyrrole derivatives, rhodium-catalyzed Suzuki-type cross-coupling and copper-catalyzed asymmetric conjugate reduction of coumarins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as a reactant for Suzuki-Miyaura cross-coupling reactions, condensation reactions, synthesis of azobenzene-functionalized compounds. Also used for preparation of highly fluorescent diketopyrrolopyrrole derivatives, rhodium-catalyzed Suzuki-type cross-coupling and copper-catalyzed asymmetric conjugate reduction of coumarins.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text