5-Methylpyridine-3-boronic acid, 98%, Thermo Scientific Chemicals
5-Methylpyridine-3-boronic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

5-Methylpyridine-3-boronic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H52891.03
also known as H52891-03
Price (USD)/ Each
215.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
215.00
Add to cart
5-Methylpyridine-3-boronic acid, 98%, Thermo Scientific Chemicals
Catalog numberH52891.03
Price (USD)/ Each
215.00
-
Add to cart
Chemical Identifiers
CAS173999-18-3
IUPAC Name(5-methylpyridin-3-yl)boronic acid
Molecular FormulaC6H8BNO2
InChI KeyREONQWGHSQHTAC-UHFFFAOYSA-N
SMILESCC1=CN=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Assay (HPLC)≥97.5%
Proton NMRConforms
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only