Thermo Scientific Chemicals

3,5-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H52965.06
5 g, Each
Thermo Scientific Chemicals

3,5-Dimethoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H52965.06
5 g, Each
Quantity
Catalog number: H52965.06
also known as H52965-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
192182-54-0
IUPAC Name
(3,5-dimethoxyphenyl)boronic acid
Molecular Formula
C8H11BO4
InChI Key
XUIURRYWQBBCCK-UHFFFAOYSA-N
SMILES
COC1=CC(=CC(OC)=C1)B(O)O
Assay (HPLC)
>97.5%

Description

3,5-Dimethoxybenzeneboronic acid is used as reactant for palladium-catalyzed coupling reactions, preparation of benzopyranone derivatives as positive GABAA receptor modulators, preparation of aryl alkenes via three-component coupling catalyzed by palladium and Suzuki-Miyaura coupling. It is also used as reactant for rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide and preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethoxybenzeneboronic acid is used as reactant for palladium-catalyzed coupling reactions, preparation of benzopyranone derivatives as positive GABAA receptor modulators, preparation of aryl alkenes via three-component coupling catalyzed by palladium and Suzuki-Miyaura coupling. It is also used as reactant for rhodium catalyzed cyanation with N-cyano-N-phenyl-p-methylbenzenesulfonamide and preparation of bisphosphonate inhibitors of human farnesyl pyrophosphate synthase.

Solubility
Soluble in methanol.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text