3-(2-Carboxyethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
3-(2-Carboxyethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-(2-Carboxyethyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H52977.03
also known as H52977-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Specifications
Chemical Name or Material3-(2-Carboxyethyl)benzeneboronic acid
CAS693803-17-7
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only