3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number H53084.03
also known as H53084-03
Price (USD)/ Each
531.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
531.00
Add to cart
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals
Catalog numberH53084.03
Price (USD)/ Each
531.00
-
Add to cart
Chemical Identifiers
CAS871329-62-3
IUPAC Name[3-fluoro-5-(methoxycarbonyl)phenyl]boronic acid
Molecular FormulaC8H8BFO4
InChI KeyVKHJVASTGLPBBL-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(F)=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>94.0%
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-5-(methoxycarbonyl)benzeneboronic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only