4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H53101.03
also known as H53101-03
Price (USD)/ Each
423.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
423.00
Add to cart
4-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
Catalog numberH53101.03
Price (USD)/ Each
423.00
-
Add to cart
Chemical Identifiers
CAS874219-19-9
IUPAC Name[4-fluoro-3-(methylcarbamoyl)phenyl]boronic acid
Molecular FormulaC8H9BFNO3
InChI KeyDVDSMLPVTUSIFM-UHFFFAOYSA-N
SMILESCNC(=O)C1=C(F)C=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>95.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only