3-Carbamoyl-4-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
3-Carbamoyl-4-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Carbamoyl-4-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
250 mg
Catalog number H53102.03
also known as H53102-03
Price (USD)/ Each
379.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
379.00
Add to cart
3-Carbamoyl-4-fluorobenzeneboronic acid, 97%, Thermo Scientific Chemicals
Catalog numberH53102.03
Price (USD)/ Each
379.00
-
Add to cart
Chemical Identifiers
CAS874219-34-8
IUPAC Name(3-carbamoyl-4-fluorophenyl)boronic acid
Molecular FormulaC7H7BFNO3
InChI KeyRNNYXSWJFLHIRC-UHFFFAOYSA-N
SMILESNC(=O)C1=C(F)C=CC(=C1)B(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only