Thermo Scientific Chemicals

3-Fluoropyridine-4-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H53109.MD
250 mg, Each
Thermo Scientific Chemicals

3-Fluoropyridine-4-boronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H53109.MD
250 mg, Each
Quantity
Catalog number: H53109.MD
also known as H53109-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
458532-97-3
IUPAC Name
(3-fluoropyridin-4-yl)boronic acid
Molecular Formula
C5H5BFNO2
InChI Key
QUHSESYLMZVXCN-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=NC=C1F
Assay (HPLC)
>97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text