Thermo Scientific Chemicals

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H53139.03
1 g, Each
Thermo Scientific Chemicals

1-Methyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog number: H53139.03
1 g, Each
Quantity
Catalog number: H53139.03
also known as H53139-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
761446-44-0
IUPAC Name
1-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Molecular Formula
C10H17BN2O2
InChI Key
UCNGGGYMLHAMJG-UHFFFAOYSA-N
SMILES
CN1C=C(C=N1)B1OC(C)(C)C(C)(C)O1
Assay (GC)
96.0% min

Description

It is a reagent used for Suzuki-Miyaura cross-coupling reactions, transesterification reactions. It is involved in several reactions as a reagent for the preparation of aminothiazoles as γ-secretase modulators,amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy, pyridine derivatives as TGF-β1 and active A signaling inhibitors and MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a reagent used for Suzuki-Miyaura cross-coupling reactions, transesterification reactions. It is involved in several reactions as a reagent for the preparation of aminothiazoles as γ-secretase modulators,amino-pyrido-indol-carboxamides, as potential JAK2 inhibitors for myeloproliferative disorders therapy, pyridine derivatives as TGF-β1 and active A signaling inhibitors and MK-2461 analogs as inhibitors of c-Met kinase for the treatment of cancer.

Solubility
Slightly soluble in water.

Notes
Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text