Thermo Scientific Chemicals

2-Phenoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H53264.06
5 g, Each
Thermo Scientific Chemicals

2-Phenoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Catalog number: H53264.06
5 g, Each
Quantity
Catalog number: H53264.06
also known as H53264-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
108238-09-1
IUPAC Name
(2-phenoxyphenyl)boronic acid
Molecular Formula
C12H11BO3
InChI Key
AVOWPOFIQZSVGV-UHFFFAOYSA-N
SMILES
OB(O)C1=CC=CC=C1OC1=CC=CC=C1
Assay (HPLC)
≥97.5%
Appearance (Color)
White
Form
Powder
Proton NMR
≥97.5% ¹H nmr

Description

Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant for palladium catalyzed cross-coupling reactions, trifluoromethylation via copper-mediated oxidative cross-coupling, preparation of biologically and pharmacologically active molecules, preparation of pyridazine-based scaffolds as a-helix mimetics.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text