Thermo Scientific Chemicals

3-(Aminomethyl)benzeneboronic acid hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H53284.MD
250 mg, Each
Thermo Scientific Chemicals

3-(Aminomethyl)benzeneboronic acid hydrochloride, 96%, Thermo Scientific Chemicals

Catalog number: H53284.MD
250 mg, Each
Quantity
Catalog number: H53284.MD
also known as H53284-MD
Price (USD)
Quantity
-
Have Questions?

Description

3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Aminomethyl)benzeneboronic acid hydrochloride is used as intermediates. Hydroxybenzene boronic acids are involved in suzuki miyaura reactions. It is a arylboronic acid. 3-Fluoro-4-hydroxybenzeneboronic acid, can be an effective catalyst for amidation and esterification of carboxylic acids.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerate.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text