4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Carboxy-3-fluorobenzeneboronic acid is used as intermediates. Also used in the coupling reaction.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
RUO – Research Use Only
Green, I. R.; et al. 2-Arylnaphthoquinone analogues: potential anti-TB and pro-apoptotic agents. Journal name. 2011, 192-212.